Learn More
Medchemexpress LLC 2-Methoxycarbonylphenylboronic acid | 374538-03-1 | 179.97 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW002908
Encompass_Preferred
2-Methoxycarbonylphenylboronic acid is an intermediate that can be used in the synthesis of BLT2 agonist. It is for research use only and not sold to patients. This product is a controlled substance and may not be for sale in certain territories.
  • Intermediate for BLT2 agonist synthesis
  • Research use only
  • Solid, white to off-white appearance
  • Available in various sizes
Catalog No. 50-004-48284