Learn More
Medchemexpress LLC 2-methoxyestradiol | 362-07-2 | >98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1203350MG
Encompass_Preferred
2-Methoxyestradiol is an endogenous metabolite of 17β-estradiol used in research as an apoptosis inducer and angiogenesis inhibitor. It destabilizes microtubules and can generate reactive oxygen species; supplied as a high-purity solid for analytical and cell-based studies.
  • Endogenous estradiol metabolite with pro-apoptotic activity.
  • Inhibits angiogenesis and disrupts microtubule function.
  • High purity suitable for analytical and cell-based assays.
  • Soluble in DMSO; practically insoluble in water and ethanol.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-34003