Learn More
Medchemexpress LLC 2-methoxyestradiol | 362-07-2 | >99.5% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1203310MM 1ML
Encompass_Preferred
2-Methoxyestradiol is an endogenous metabolite of 17β-estradiol used in research to investigate apoptosis, antiproliferative, and antiangiogenic effects. Supplied as a 10 mM, 1 mL stock solution in DMSO, it is intended for use in in vitro and preclinical assays studying microtubule disruption and angiogenesis.
  • High purity suitable for research-grade experiments.
  • Supplied as a ready-to-use 10 mM solution in DMSO for immediate assay use.
  • Appropriate for in vitro cell biology and preclinical antiangiogenesis studies.
  • Stable when stored frozen; follow storage recommendations to avoid degradation.
Catalog No. 50-002-31700