Learn More
Medchemexpress LLC 2-methoxyestradiol | 362-07-2 | 99.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY12033100MG
Encompass_Preferred
2-methoxyestradiol is an endogenous metabolite of 17β-estradiol used as a research reagent with apoptosis-inducing and antiangiogenic activity. Supplied as a high-purity solid for analytical and biological research applications.
  • High purity suitable for analytical work
  • Available in research quantities, including 100 MG
  • Used in apoptosis and antiangiogenesis studies
  • Soluble in DMSO for solution preparation
  • Intended for research use only
Catalog No. 50-002-34210