Learn More
Medchemexpress LLC 2-methoxyidazoxan monohydrochloride | 109544-45-8 | MFCD00069343 | 98.6% | 270.71 | C12H15ClN2O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1031975MG
Encompass_Preferred
2-Methoxyidazoxan monohydrochloride is a research reagent that functions as a selective alpha-2 adrenoceptor antagonist with minimal imidazoline activity. Supplied as a white to off-white solid, it is used in pharmacological studies to probe alpha-2 adrenergic receptor subtype function and shows higher affinity for alpha-2D than alpha-2A receptors.
  • Selective alpha-2 adrenoceptor antagonist for receptor pharmacology studies.
  • Minimal imidazoline antagonist activity.
  • Molecular formula C12H15ClN2O3; molecular weight 270.71 g/mol.
  • High purity (98.6%).
  • White to off-white solid, suitable for formulation or dissolution in common solvents.
Catalog No. 50-002-47271