Learn More
Medchemexpress LLC 2-methoxyidazoxan monohydrochloride | 109544-45-8 | MFCD00069343 | 98.6% | 270.71 g/mol | C12H15ClN2O3 | 100 MG

Supplier: Medchemexpress LLC HY103197100MG
2-Methoxyidazoxan monohydrochloride is a research reagent and selective alpha-2 adrenoceptor antagonist supplied as the monohydrochloride salt for pharmacology and receptor-binding studies. It is provided as a high-purity solid with defined solubility and storage recommendations.
- High purity: 98.64% (reported by supplier).
- Molecular formula: C12H15ClN2O3; molecular weight: 270.71 g/mol.
- Soluble in water: 100 mg/mL (requires ultrasonic); in PBS: 50 mg/mL (clear solution).
- Available in multiple pack sizes, including 100 MG.
- Recommended storage: solid at 4°C under nitrogen; in solution -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.