Learn More
Medchemexpress LLC 2-Methoxyphenothiazine | 1771-18-2 | 99.6% | 229.30 | 50 G
Supplier: Medchemexpress LLC HYW01225850G
2-Methoxyphenothiazine (2-Methoxy-10H-phenothiazine) is a biochemical reagent that can be used in research fields such as life sciences or organic chemistry.
- For research use only.
- Enhances decolorization efficiency of laccase from Trametes modesta towards various dyes.
- Functions as an active component of a catalyst for electrochemically intensified esterification.
- Exhibits off-white to gray appearance.
- Inhibits erastin-induced ferroptosis in human HT-1080 cells.
- Inhibits 5-lipoxygenase pathway in rat basophilic leukemia (RBL-1) cells.