Learn More
Medchemexpress LLC 2-methyl-1,3-cyclohexanedione | 1193-55-1 | MFCD00001587 | 99.9% | 126.15 g/mol | C7H10O2 | 50 G

Supplier: Medchemexpress LLC HYW01877850G
2-Methyl-1,3-cyclohexanedione is a small-molecule organic compound used as a synthetic intermediate in pharmaceutical and specialty-chemical synthesis. It is supplied as a solid with high analytical purity for research and development and is reported for use in cosmetic ingredient applications.
- High purity (≈99.9%).
- Molecular formula C7H10O2.
- Molecular weight 126.15 g/mol.
- Solid form suitable for handling and storage.
- Useful synthetic intermediate for drug and specialty-chemical synthesis.
- Available in multiple laboratory pack sizes for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.