Learn More
Medchemexpress LLC 2-Methyl-2-(4-(2-methyl-8-(1H-pyrrolo[2,3-b]pyridin-6-yl)-1H-naphtho[1,2-d]imidazol-1-yl)phenyl)p... | 2845104-25-6 | 99.7% | 443.50 g·mol⁻¹ | C27H21N7 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15162210MG
Encompass_Preferred
An orally active small-molecule dual inhibitor of PI3K and mTOR developed for preclinical cancer research. Supplied as a high-purity solid with recommended solubility and storage conditions for both in vitro and in vivo experiments.
  • Dual PI3K and mTOR inhibition with low-nanomolar potency (PI3Kα 3.5 nM; PI3Kδ 4.6 nM; mTOR 21.3 nM)
  • High purity suitable for research applications (99.65%)
  • Solid, off-white to gray appearance and stable under recommended storage
  • Soluble in DMSO (40 mg/mL) and formulatable for in vivo dosing (10% DMSO/corn oil)
  • Multiple packaging formats for in vitro and in vivo use
  • Includes storage guidance for solutions (-80°C/-20°C) and solids (4°C protected from light)
Catalog No. 50-002-87419