Learn More
eMolecules​ 1115492-92-6 | 2-Methyl-4-pyridylzinc bromide, 0.25 M in THF | Synthonix237.410 | C6H6BrNZn | 0.000 | [Br-].Cc1cc([Zn+])ccn1 | 25ml | 627353690
Supplier:  eMolecules​ 627353690
Encompass_Preferred
2-Methyl-4-pyridylzinc bromide, 0.25 M in THF | Synthonix | 1115492-92-6237.410 | C6H6BrNZn | 0.000 | [Br-].Cc1cc([Zn+])ccn1 | 25ml | 627353690
Catalog No. 50-269-5288