Learn More
Medchemexpress LLC 2-methyl-6-(2-phenylethenyl)pyridine | 6266-99-5 | MFCD02262120 | 99.8% | 195.26 g·mol⁻¹ | C14H13N | 1 ML

Supplier: Medchemexpress LLC HY10209410MM 1ML
(E/Z)-SIB-1893 is a racemic mixture of the (E)- and (Z)-isomers of SIB-1893, a selective non-competitive antagonist of the metabotropic glutamate receptor subtype 5 (mGluR5). It is supplied as a 10 mM solution in DMSO for research use.
- Racemic mixture of (E)- and (Z)-isomers.
- Selective non-competitive mGluR5 antagonist.
- Supplied as 10 mM solution in DMSO, 1 mL vial.
- Purity 99.8%.
- Molecular weight 195.26 g·mol⁻¹; formula C14H13N.
- Store per manufacturer recommendations: powder -20°C (3 years) or 4°C (2 years).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.