Learn More
Medchemexpress LLC 2-(methylamino)benzoic acid | 119-68-6 | MFCD02668743 | 99.9% | 151.16 g/mol | C8H9NO2 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW040705100G
Encompass_Preferred
2-(Methylamino)benzoic acid (N-methylanthranilic acid) is a research-grade small molecule and a primary metabolite of methyl-N-methylanthranilates provided for laboratory and analytical use; it is not intended for human, clinical, or food applications.
  • High purity reported at 99.93%.
  • Molecular formula C8H9NO2 and molecular weight 151.16 g/mol.
  • Available in common laboratory package sizes, including 100 G.
  • Suitable as a metabolite reference and analytical standard for research applications.
Catalog No. 50-002-08513