Learn More
Medchemexpress LLC 2-methylbutanal | 96-17-3 | MFCD00006984 | 98.6% | 86.13 g/mol | C5H10O | 100 G
SDP
Supplier:  Medchemexpress LLC HYW020678100G
Encompass_Preferred
2-Methylbutanal (2-methylbutyraldehyde) is a volatile aliphatic aldehyde supplied for laboratory research and analytical use. It is reported as a biomarker for detection of the uropathogen Proteus and is provided as a liquid reagent with reported high purity suitable for chromatographic and biochemical applications.
  • Volatile aldehyde used as an analytical standard.
  • Reported biomarker for detection of Proteus species.
  • High reported purity (98.56%).
  • Liquid physical state; density approximately 0.8131 g/cm³.
  • Suitable for chromatographic and analytical workflows.
Catalog No. 50-003-00742