Learn More
Medchemexpress LLC 2-methylcitric acid | 6061-96-7 | MFCD01074482 | 99.7% | 206.15 g/mol | C7H10O7 | 5 MG

Supplier: Medchemexpress LLC HY1133715MG
2-Methylcitric acid is an endogenous tricarboxylic acid supplied as a research-grade chemical standard for analytical and biochemical applications. It is provided as a white to off-white solid and as a DMSO solution for method development, metabolite research, and LC-MS/MS workflows.
- High purity (99.72%).
- Molecular weight 206.15 g/mol.
- Chemical formula C7H10O7.
- Available as solid (5 mg, 10 mg, 25 mg) and as 10 mM solution in DMSO.
- Suitable for analytical reference, method validation, and pathway studies.
- Documentation available: datasheet, certificate of analysis, and SDS.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.