Learn More
Medchemexpress LLC 2-Methylcitric acid trisodium | 117041-96-0 | 96.0% | 272.10 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY113371A10MG
Encompass_Preferred
2-Methylcitric acid trisodium is an endogenous metabolite involved in the 2-methylcitric acid cycle. It accumulates in methylmalonic and propionic acidemias, serving as a marker metabolite. This compound significantly inhibits ADP-stimulated and uncoupled respiration in mitochondria supported by glutamate.
  • Accumulates in methylmalonic and propionic acidemias
  • Acts as a marker metabolite
  • Inhibits ADP-stimulated and uncoupled mitochondrial respiration
Catalog No. 50-004-37053