Learn More
Medchemexpress LLC 2-Methylcyclohexanone | 583-60-8 | 98.9% | 112.17 | 100 G

Supplier: Medchemexpress LLC HYW010549100G
2-Methylcyclohexanone is a chemical compound, appearing as a colorless to light yellow liquid. With a molecular weight of 112.17 and a molecular formula of C7H12O, this substance is suitable for various research applications.
- Consistent with structure as confirmed by 1H NMR spectrum
- High purity (GC) of 98.89%
- Low water content (KF) of 0.18%
- Long-term storage stability in pure form at -20°C for 3 years or 4°C for 2 years
- Stable in solvent when stored at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.