Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Methylhexanoic acid | 4536-23-6 | 98.0% | 500 MG
SDP

Supplier:  Medchemexpress LLC HY128371500MG

Encompass_Preferred

2-methylhexanoic acid is a medium-chain fatty acid primarily used as a flavoring agent. This product is a colorless to light yellow liquid with a molecular weight of 130.19, and a chemical formula of C7H14O2.

  • Medium-chain fatty acid
  • Functions as a flavoring agent
  • Colorless to light yellow liquid appearance
  • Molecular weight of 130.19
  • Chemical formula: C7H14O2
  • Purity of 98.0%

Catalog No. 50-003-39758


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.