Learn More
Cayman Chemical 2-MethylhIppurIc AcId 1g
SDP
Supplier:  Cayman Chemical 343761 G
Encompass_Preferred
A metabolite of xylene; has been found in the urine of industrial workers exposed to xylene
Catalog No. 50-294-9913