Learn More
ALADDIN SCIENTIFIC CORPORATION 2-Methylimidazole, 250g
Supplier:  ALADDIN SCIENTIFIC CORPORATION M104839250G
Encompass_Preferred
2-Methylimidazole is used as a raw material for the preparation of nitroimidazole antibiotics which is useful in combat anaerobic bacterial and parasitic infections It acts as a ligand in coordination chemistry It is also employed as a hardener or accelerator for epoxy resin and auxiliary agent for textile dyes Further it acts as a catalyst for refolding of enhanced colored fluorescent protein It is also useful as a building block in the synthesis of biologically active compounds In addition to this it is used as an active pharmaceutical ingredient intermediate such as metronidazole and dimetridazole
Catalog No. 50-000-82572