Learn More
Medchemexpress LLC 2-Methylindole | 95-20-5 | 131.17 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW007556100G
Encompass_Preferred
2-Methylindole is a biochemical reagent used as a biological material or organic compound for life science-related research. It serves as a reactant in various synthesis reactions, including regioselective synthesis of oxopyrrolidine analogs, Friedel-Crafts alkylation reactions, and the preparation of tryptophan dioxygenase inhibitors. It is also utilized in Michael addition reactions and the synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.
  • Biochemical reagent for life science research
  • Used as a biological material or organic compound
  • Reactant for regioselective synthesis of oxopyrrolidine analogs
  • Reactant for Friedel-Crafts alkylation reactions
  • Reactant for preparation of tryptophan dioxygenase inhibitors
  • Reactant for Michael addition reactions
  • Reactant for synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors
Catalog No. 50-004-47550