Learn More
Medchemexpress LLC 2-Methylpropylboronic acid | 84110-40-7 | 101.94 | 25 G

Supplier: Medchemexpress LLC 25GHYW000978
(2-Methylpropyl)boronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It appears as a white to off-white solid. This product is for research use only.
- Appearance: Solid, white to off-white
- Formula: C4H11BO2
- Shipping: Room temperature in continental US
- Storage (powder): -20°C for 3 years
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month
- Solubility (in vitro): DMSO: 200 mg/mL (1961.94 mM), requires ultrasonic assistance
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.