Learn More
Medchemexpress LLC 2-Methyltetrahydrofuran-3-one | 3188-00-9 | MFCD00010423 | 99.7% | 100.12 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01060810MM/1ML
Encompass_Preferred
2-Methyltetrahydrofuran-3-one is identified as a quorum sensing inhibitor capable of preventing biofilm formation by *H. alvei*. It is also recognized as one of the volatile components found in roasted coffee. This product is intended for research use only and is not sold to patients.
  • Quorum sensing inhibitor
  • Inhibits biofilm formation by *H. alvei*
  • One of the volatile constituents of roasted coffee
Catalog No. 50-004-53944