Learn More
Sigma Aldrich Fine Chemicals Biosciences 2 Methyltetrahydrofuran Bi200L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 155810200L
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. This product is a biorenewable and thus aligns with Safer Solvents and Auxiliaries and Use of Renewable feedstocks. Click here for more information.2-Methyltetrahydrofuran is an environmentally favorable alternative to tetrahydrofuran (THf) 1 4 Dioxane (Dioxane) and dichloromethane (DCM) for most industrial applications. Many of its benefits include High boiling pointClean organic-water phase separationReduced energy to recover
Catalog No. 50-290-3836
Only null left
Add to Cart
Request a Quote