Learn More
Eurofins Discoverx 2-Methylthio-ATP, 10 mg. (92-1214)
Supplier:  Eurofins Discoverx 921214
Encompass
2-(Methylthio)adenosine 5′-triphosphate (2-methylthio-ATP) is an ATP analog that is a more selective agonist of the P2γ purinergic receptor. Adenosine 5'-triphosphate is a nucleoside triphosphate that is key to intracellular energy transfer. It is responsible for transporting chemical energy within cells for metabolism, is an end product of phosphorylation, cellular respiration, and fermentation, and is used in many cellular processes. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9739