Learn More
Cayman Chemical 2-MIcroglobulIndog Moncl 100ug
SDP
Supplier:  Cayman Chemical 31017100 UG
Encompass_Preferred
Immunogen: Recombinant dog β2-microglobulin protein (amino acids 28-125) • Host: Mouse • Species Reactivity: (+) Dog; (-) Human • Cross Reactivity: (+) β2-microglobulin • Applications: FC, IP, WB • MW = ~14 kDa
Catalog No. 50-299-1425