Learn More
Abcam 2 mm Coronal Mouse Brain Slicer, 1UNIT
Supplier:  Abcam AB2433121UNIT
Encompass_Preferred
This 2 mm coronal mouse brain slicer allows you to easily cut a brain into smaller pieces for easier imaging with objectives that have a limited working distance.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2466