Learn More
Medchemexpress LLC 2-morpholinoethanesulfonic acid-d13 | 352534-94-2 | 98.8% | 208.32 g/mol | C6D13NO4S | 10 MG

Supplier: Medchemexpress LLC HYD0858S10MG
MES-d13 is the deuterium-labeled form of 2-morpholinoethanesulfonic acid (MES), provided as a stable-isotope reagent for analytical and research use. It is commonly used as an internal standard and buffering agent in NMR and LC-MS workflows requiring isotopic labeling.
- Deuteration level d13.
- Molecular formula C6D13NO4S.
- Molecular weight 208.32 g/mol.
- Purity 98.8% (as listed).
- Available in milligram quantities, including a 10 mg vial.
- Recommended storage: powder -20°C for long term; in solvent -80°C for up to six months.
- Applications: NMR, LC-MS, and isotopic internal standard use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.