Learn More
Medchemexpress LLC 2-(Morpholinothio)benzothiazole | 102-77-2 | MFCD00022870 | 99.2% | 252.36 | 250 G

Supplier: Medchemexpress LLC 250GHY139432
2-(Morpholinothio)benzothiazole is an accelerator used for amperometric measurements. This solid appears as light yellow to yellow in color. It is for research use only and not sold to patients.
- Purity of 99.21%
- Molecular weight of 252.36
- Chemical formula C11H12N2OS2
- Solid appearance
- Light yellow to yellow color
- Ships at room temperature in continental US
- Store at 4°C under nitrogen
- In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
- Soluble in DMSO at 50 mg/mL
- Functions as a delayed-action vulcanization accelerator
- Insoluble in water, soluble in benzene, acetone, and methanol
- Melting point of 80 °C
- Flash point of 188 °C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.