Learn More
Medchemexpress LLC 2-(naphthalen-2-yl)acetonitrile | 7498-57-9 | MFCD00004122 | 100.0% | 167.21 g·mol⁻¹ | C12H9N | 5 G

Supplier: Medchemexpress LLC HYW0171165G
2-(Naphthalen-2-yl)acetonitrile is a small-molecule intermediate used primarily in medicinal chemistry and organic synthesis. It serves as a building block for preparing more complex target molecules in research and development and is supplied with characterization and storage guidance for laboratory use.
- Solid, white to off-white appearance.
- High purity suitable for synthetic workflows.
- Molecular formula C12H9N; molecular weight 167.21 g·mol⁻¹.
- CAS number 7498-57-9 for unambiguous identification.
- Intended for use as a chemical intermediate in research and development.
- Store at room temperature; in solution, keep at -20°C to -80°C as recommended.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.