Learn More
Medchemexpress LLC 2-Naphthaleneacetic acid, 6-methoxy-α-methyl- | 23981-80-8 | C14H14O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1502910MM/1ML
Encompass_Preferred
(±)-Naproxen is a racemate of Naproxen, functioning as a COX-1 and COX-2 inhibitor. It exhibits IC50s of 8.72 μM for COX-1 and 5.15 μM for COX-2.
  • High purity of 99.79%
  • White to light yellow solid appearance
  • Molecular weight of 230.26
  • Readily soluble in DMSO
  • Available as a 10 mM solution in DMSO
  • Specific storage conditions for powder and solvent forms
Catalog No. 50-003-75334