Learn More
Medchemexpress LLC 2-Naphthalenemethanol | 1592-38-7 | MFCD00004124 | 158.20 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0019535G
Encompass_Preferred
2-Naphthalenemethanol is a metabolite of the environmental pollutant 2-methylnaphthalene. It covalently binds to alveolar protein and induces pulmonary toxicity. It appears as a white to off-white solid with the molecular formula C11H10O.
  • Metabolite of environmental pollutant 2-methylnaphthalene
  • Covalently binds to alveolar protein
  • Induces pulmonary toxicity
  • White to off-white solid
  • Molecular formula: C11H10O
Catalog No. 50-004-47928