Learn More
Medchemexpress LLC 2-Naphthalenol, 1-[4,5-dihydro-5-(4-methoxyphenyl)-1H-pyrazol-3-yl]- | 152367-54-9 | 99.6% | 318.37 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY169118
Encompass_Preferred
EGR-1-IN-2 (compound 2) is an EGR-1 inhibitor with anti-inflammatory activity. It inhibits the formation of EGR-1-DNA complexes induced by TNF-α in HaCaT cells. This product is for research use only.
  • Inhibits EGR-1-DNA complexes
  • Anti-inflammatory activity
  • For research use only
Catalog No. 50-003-94051