Learn More
Medchemexpress LLC 2-naphthol | 135-19-3 | MFCD00004067 | 99.9% | 144.17 g/mol | C10H8O | 500g
SDP
Supplier:  Medchemexpress LLC HYY0110500G
Encompass_Preferred
2-Naphthol is a metabolite of naphthalene catalyzed by cytochrome P450 (CYP) isozymes (CYP 1A1 CYP 1A2 CYP 2A1 CYP 2E1 and CYP 2F2)
Catalog No. 50-002-69025