Learn More
Medchemexpress LLC 2-naphthoxyacetic acid | 120-23-0 | MFCD00004066 | 99.9% | 202.21 g/mol | C12H10O3 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01479650G
Encompass_Preferred
2-Naphthoxyacetic acid is a small aromatic carboxylic acid used as a research-grade endogenous metabolite standard and reagent. It is supplied as a solid and as DMSO solutions for biochemical, analytical, and metabolic studies.
  • High purity (99.89%) suitable for analytical applications.
  • Molecular weight 202.21 g/mol and formula C12H10O3.
  • Available in multiple pack sizes to support bench-scale and bulk needs.
  • White to off-white solid with good solubility in DMSO.
  • Commonly used as a reference standard and in metabolite profiling workflows.
Catalog No. 50-003-02886