Learn More
Cayman Chemical 2-NBDG, 186689-07-6, 5 mg
SDP
Supplier:  Cayman Chemical 110465
Encompass_Preferred
A fluorescent derivative of glucose; uptake is competitively inhibited by D-glucose in E. coli; has been used to monitor glucose uptake by bacteria and live mammalian cells and in tumor biopsies; ex/em = 475/550 nm, respectively,
Catalog No. NC0292035