Learn More
Carbosynth LLC. 2-NBDG (0.5 mg vial), 1ea, MW: 342.26, Molecular formula: C12H14N4O8, CAS# 186689-07-6
SDP
Supplier:  Carbosynth LLC. CKG23002V0.5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: 2-[N-(7-Nitrobenz-2-Oxa-1,3-Diazol-4-yl)Amino]-2- Deoxy-D-Glucose, Application: Reagent for Monitoring Glucose Uptake Reagent for Monitoring Glucose Uptake into Single, Living Cells, Storage Temperature: -20C, References: K. Yoshioka, H. Takahashi, T. Homma, M. Saito, K.-B. Oh, Y. Nemoto, and H. Matsuoka, Biochim. Biophys. Acta, 1289, 5 (1996). (Original), K. Yamada, M. Nakata, N. Horimoto, M. Saito, H. Matsuoka, and N. Inagaki, J. Biol. Chem., 275, 22278 (2000). (Measurement of Glucose Uptake in Living Mammalian Cells), K. Yamada, M. Saito, H. Matsuoka, and N. Inagaki, Nature Protocols, 2, 1 (2007). (Chem. Synthesis & Protocols for Measurement).
Catalog No. 50-168-5737