Learn More
Medchemexpress LLC 2-nitrobenzamide | 610-15-1 | MFCD00007976 | 99.2% | 166.13 g/mol | C7H6N2O3 | 50 G

Supplier: Medchemexpress LLC HYW02765450G
2-nitrobenzamide (CAS 610-15-1) is an organic intermediate commonly used in the synthesis of pharmaceuticals and specialty organic compounds. It is supplied as a solid with reported high purity and is intended for research and small-scale synthetic applications.
- High chemical purity suitable for research (reported HPLC purity 99.16%).
- Useful building block for synthesis of active compounds and intermediates.
- Solid form simplifies handling and accurate weighing for bench-scale reactions.
- Molecular formula C7H6N2O3 and molecular weight 166.13 g/mol.
- Available in multiple lab-scale pack sizes, including 50 g, to support different experimental needs.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.