Learn More
Medchemexpress LLC 2-nitrobenzyl alcohol | 612-25-9 | MFCD00007186 | 100.0% | 153.14 g/mol | C7H7NO3 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW015569
Encompass_Preferred
2-Nitrobenzyl alcohol is an organic reagent used as a synthetic intermediate and for photolabile protecting-group chemistry in chemical and life-science research. It is provided as an off-white to light brown solid with analytical data supporting suitability for research use.
  • High purity suitable for research (99.96% by HPLC).
  • Molecular formula C7H7NO3 and molecular weight 153.14 g/mol.
  • Off-white to light brown solid, easy to handle and store.
  • Useful as a building block and in protecting-group chemistry.
  • Available in convenient laboratory pack sizes for small-scale research.
Catalog No. 50-003-03085