Learn More
Alkali Scientific 2 Nitrophenyl β D Xylopyranoside, 500 Milligrams
SDP
Supplier:  Alkali Scientific ENY001CP
Encompass_Preferred
2-Nitrophenyl β-D-xylopyranoside (500 mg) is a substrate used for studying xylanase activity in enzymatic assays. Xylanases, which break down xylan (a hemicellulose), can cleave this substrate, producing p-nitrophenol that is easily measured by its absorbance at 405 nm. This reagent is crucial for biochemists studying the hydrolysis of hemicellulose, a major component of plant cell walls. It has applications in areas such as agricultural research, biofuel production, and the characterization of enzymes that degrade plant biomass. The 500 mg quantity makes it suitable for both small- and large-scale enzyme activity testing.
Catalog No. 50-006-59886