Learn More
Cayman Chemical 2-NonylquInolIn-4 1H-on 10mg
SDP
Supplier:  Cayman Chemical 900362710 MG
Encompass_Preferred
A quinolone alkaloid; active against HCV (IC50 = 1.4 UG/ml) and L. gongylophorus; inhibits TPA- and A23187-induced NFAT activation in Jurkat cells but not LPS-induced NF-κB activation in RAW 264.7 cells (IC50s = 3.44 and >100 µM, respectively, in reporter assays)
Catalog No. 50-289-5904