Learn More
Cayman Chemical 2-NonylquInolIn-4 1H-on-d4 1mg
SDP
Supplier:  Cayman Chemical 90039701 MG
Encompass_Preferred
An internal standard for the quantification of 2-nonylquinolin-4(1H)-one by GC- or LC-MS
Catalog No. 50-289-5910