Learn More
Medchemexpress LLC 2'-o-(2-methoxyethyl)adenosine | 168427-74-5 | MFCD02682961 | 99.0% | 325.32 | C13H19N5O5 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW048491
2′-O-(2-Methoxyethyl)adenosine is a 2′-O-methoxyethyl-modified adenosine nucleoside used as a building block in oligonucleotide synthesis and related research applications. It is supplied as a solid and as a DMSO solution, and is provided at high purity for research use.
- Used as a building block for oligonucleotide synthesis.
- Available as solid and as a 10 mM solution in DMSO.
- High purity (99.0%).
- Molecular weight 325.32 and formula C13H19N5O5.
- CAS number 168427-74-5 for chemical identification.
- Intended for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.