Learn More
Medchemexpress LLC 2'-O-(2-Methoxyethyl) inosine | 545374-76-3 | 973934 | C13H18N4O6 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY152516
Encompass_Preferred
2'-O-(2-Methoxyethyl) inosine is a purine nucleoside analog known for its broad antitumor activity, especially against indolent lymphoid malignancies. Its mechanisms of action involve the inhibition of DNA synthesis and the induction of apoptosis.
  • Purine nucleoside analog
  • Broad antitumor activity
  • Effective against indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
Catalog No. 50-003-74760