Learn More
Medchemexpress LLC 2'-O-(2-Methoxyethyl) inosine | 545374-76-3 | C13H18N4O6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15251650MG
Encompass_Preferred
2'-O-(2-Methoxyethyl) inosine is a purine nucleoside analog that exhibits broad antitumor activity, particularly against indolent lymphoid malignancies. Its anticancer mechanisms involve inhibiting DNA synthesis and inducing apoptosis.
  • Purine nucleoside analog
  • Exhibits broad antitumor activity
  • Effective against indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
Catalog No. 50-003-73054