Learn More
Medchemexpress LLC 2'-O,4'-C-methylenecytidine | 206055-69-8 | MFCD28342628 | 98.6% | 255.23 | C10H13N3O5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW57088810MM/1ML
Encompass_Preferred
2'-O,4'-C-Methylenecytidine is a bicyclic locked nucleic acid (LNA) cytidine analogue used to enhance duplex stability in oligonucleotide research. It is supplied as a ready-to-use 10 mM solution in DMSO or as a solid; follow recommended storage and handling for solvent and solid forms.
  • Stabilizes duplex formation with complementary DNA and RNA.
  • Suitable for incorporation into oligonucleotides.
  • High purity, approximately 98.6%.
  • Available as 10 mM solution in DMSO or as a solid.
  • Molecular weight 255.23 g/mol.
  • Intended for research use only; not for clinical use.
Catalog No. 50-002-84111