Learn More
Medchemexpress LLC 2'-O,4'-C-methyleneguanosine | 207131-16-6 | MFCD31707476 | 99.2% | 295.25 | C11H13N5O5 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW406070250MG
Encompass_Preferred
2'-O,4'-C-Methyleneguanosine is a locked nucleic acid (LNA) guanosine analogue used as a research reagent for oligonucleotide modification and nucleic acid chemistry. It is supplied as a high-purity chemical for laboratory use and is accompanied by documentation for quality and safe handling.
  • Locked nucleic acid guanosine analogue for oligonucleotide modification.
  • High chemical purity (99.2%).
  • Suitable for nucleic acid chemistry and research applications (non-clinical).
  • Available in multiple pack sizes for laboratory use.
  • Certificate of analysis and safety data sheet available.
Catalog No. 50-002-81667