Learn More
Cayman Chemical 2-O-methyl PAF C-18hydrate 5mg
SDP
Supplier:  Cayman Chemical 609125 MG
Encompass_Preferred
A synthetic PAF analog which contains an ether-linked methyl group at the sn-2 position; decreases plasma membrane fluidity, inhibits tumor cell invasiveness, and causes the release of large quantities of NO by a pathway involving activation of NOS
Catalog No. 50-289-5259