Learn More
Cayman Chemical 2-O-MethyladenosIn 10mg
SDP
Supplier:  Cayman Chemical 1693610 MG
Encompass_Preferred
An analog of adenosine used to prepare nucleoside derivatives as inhibitors of viral RNA translation and replication
Catalog No. 50-296-5885