Learn More
Cayman Chemical Astoloca mnsuns Astolon 25mg
SDP
Supplier:  Cayman Chemical 3615725 MG
Encompass_Preferred
A sesquiterpene with anticancer activity; cytotoxic to H460, ES-2, and DU145 cells (IC50s = 51.3, 61.5, and 58 UG/ml, respectively)
Catalog No. 50-291-1825