Learn More
Medchemexpress LLC 2'-OMe-A(Bz) Phosphoramidite | 110782-31-5 | 99.7% | 887.96 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13858010MM/1ML
Encompass_Preferred
2'-OMe-A(Bz) Phosphoramidite is a modified phosphoramidite monomer specifically developed for oligonucleotide synthesis. This compound facilitates the construction of diverse modified oligonucleotides, which significantly influence the properties of oligonucleotides, such as their affinity for RNA.
  • Can be used to construct various types of modified oligonucleotides.
  • Significantly impacts the properties of oligonucleotides.
  • Improves affinity for RNA.
Catalog No. 50-003-50020